MilliporeSigma
  • Role of desacetylation in the detoxification of cephalothin in renal cells in culture.
Role of desacetylation in the detoxification of cephalothin in renal cells in culture.
Journal of toxicology and environmental health (1987-01-01)
G H Hottendorf, D A Laska, P D Williams, S M Ford
ABSTRACT
The toxicity of three cephalosporin antibiotics to rabbit kidney cells in culture was compared to their known nephrotoxic potential in vivo (cephaloridine greater than cefazolin greater than cephalothin). While cephalothin is considered to be a relatively nonnephrotoxic cephalosporin when administered to many species including humans and rabbits, in several in vitro systems involving rabbit renal tissue, cephalothin was comparatively more toxic than anticipated based on in vivo data. Cephalothin is extensively desacetylated in rabbits to a less microbiologically active metabolite, desacetylcephalothin. When a microsomal S9 fraction from rabbit kidney was added to the in vitro assay in cultured rabbit renal cells, cephalothin was desacetylated and its toxicity to kidney cells was reduced. The addition of S9 in vitro provided a toxicity ranking of the cephalosporins that correlated with their known in vivo nephrotoxic potentials (cephaloridine greater than cefazolin greater than cephalothin). The in vitro detoxification of cephalothin by S9 was blocked by the coadministration of the esterase inhibitor, aminocarb. Desacetylcephalothin was relatively nontoxic to rabbit renal tissue in vitro. These results suggest that the desacetylation of cephalothin in vivo represents a previously unrecognized mechanism of detoxification of this cephalosporin antibiotic. Furthermore, this mechanism of detoxification may be applicable to other acetylated cephalosporins.
Product Description
Supelco
Aminocarb, PESTANAL®, analytical standard