MilliporeSigma
  • Interactions between filamin A and MMP-9 regulate proliferation and invasion in renal cell carcinoma.
Interactions between filamin A and MMP-9 regulate proliferation and invasion in renal cell carcinoma.
Asian Pacific journal of cancer prevention : APJCP (2014-05-30)
Guo-Gui Sun, Cui-Da Wei, Shao-Wu Jing, Wan-Ning Hu
ABSTRACT
This study aimed to analyze the expression, clinical significance of filamin A (FLNA) in renal cell carcinoma (RCC) and biological effects in a cell line by regulating FLNA expression. Immunohistochemistry and Western blotting were used to analyze FLNA protein expression in 70 cases of RCC and normal tissues to study the relationship with clinical factors. FLNA lentiviral and empty vectors were transfected into RCC to study the influence of up-regulated expression of FLNA. FLNA siRNA was transiently transfected into ACHN kidney carcinoma cells by a liposome-mediated method and protein was detected by Western blotting. The level of expression was found to be significantly lower in RCC than normal tissues (p<0.05). No correlation was noted with gender, age, tumor size or pathological types (p>0.05), but links with lymph node metastasis, clinic stage and histological grade were noted (p<0.05). Loss of FLNA expression correlated significantly with poor overall survival time by Kaplan-Meier analysis (p<0.05). Results for biological function showed that ACHN cells transfected with FLNA had a lower survival fraction, significant decrease in migration and invasion, higher cell apoptosis, higher percentage of the G0/G1 phases, and lower MMP-9 protein expression compared with ACHN cells untransfected with FLNA (p<0.05). However, renal 786-0 cells transfected with FLNA siRNA had a higher survival fraction, significant increase in migration and invasion, and higher MMP-9 protein expression compared (p<0.05). In conclusion, FLNA expression was decreased in RCC and correlated significantly with lymph node metastasis, clinic stage, histological grade and poor overall survival, suggesting that FLNA may play important roles as a a tumor suppressor in RCC by promoting degradation of MMP-9.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O