MilliporeSigma
  • Fluorouracil, epirubicin, and cyclophosphamide with either docetaxel or vinorelbine, with or without trastuzumab, as adjuvant treatments of breast cancer: final results of the FinHer Trial.
Fluorouracil, epirubicin, and cyclophosphamide with either docetaxel or vinorelbine, with or without trastuzumab, as adjuvant treatments of breast cancer: final results of the FinHer Trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2009-11-04)
Heikki Joensuu, Petri Bono, Vesa Kataja, Tuomo Alanko, Riitta Kokko, Raija Asola, Tapio Utriainen, Taina Turpeenniemi-Hujanen, Sirkku Jyrkkiö, Kari Möykkynen, Leena Helle, Seija Ingalsuo, Marjo Pajunen, Mauri Huusko, Tapio Salminen, Päivi Auvinen, Hannu Leinonen, Mika Leinonen, Jorma Isola, Pirkko-Liisa Kellokumpu-Lehtinen
ABSTRACT
Docetaxel has not been compared with vinorelbine as adjuvant treatment of early breast cancer. Efficacy and long-term safety of a short course of adjuvant trastuzumab administered concomitantly with chemotherapy for human epidermal growth factor receptor 2 (HER2) -positive cancer are unknown. One thousand ten women with axillary node-positive or high-risk node-negative breast cancer were randomly assigned to receive three cycles of docetaxel or vinorelbine, followed in both groups by three cycles of fluorouracil, epirubicin, and cyclophosphamide (FEC). Women with HER2-positive cancer (n = 232) were further assigned to either receive or not receive trastuzumab for 9 weeks with docetaxel or vinorelbine. The median follow-up time was 62 months after random assignment. Women assigned to docetaxel had better distant disease-free survival (DDFS) than those assigned to vinorelbine (hazard ratio [HR] = 0.66; 95% CI, 0.49 to 0.91; P = .010). In the subgroup of HER2-positive disease, patients treated with trastuzumab tended to have better DDFS than those treated with chemotherapy only (HR = 0.65; 95% CI, 0.38 to 1.12; P = .12; with adjustment for presence of axillary nodal metastases, HR = 0.57; P = .047). In exploratory analyses, docetaxel, trastuzumab, and FEC improved DDFS compared with docetaxel plus FEC (HR = 0.32; P = .029) and vinorelbine, trastuzumab, and FEC (HR = 0.31; P = .020). The median left ventricular ejection fraction of trastuzumab-treated patients remained unaltered during the 5-year follow-up; only one woman treated with trastuzumab was diagnosed with a heart failure. Adjuvant treatment with docetaxel improves DDFS compared with vinorelbine. A brief course of trastuzumab administered concomitantly with docetaxel is safe and effective and warrants further evaluation.