MilliporeSigma
  • Coinfection of the central nervous system by cytomegalovirus and herpes simplex virus type 1 or 2 in AIDS patients: autopsy study on 82 cases by immunohistochemistry and polymerase chain reaction.
Coinfection of the central nervous system by cytomegalovirus and herpes simplex virus type 1 or 2 in AIDS patients: autopsy study on 82 cases by immunohistochemistry and polymerase chain reaction.
Acta neuropathologica (1996-10-01)
L Vago, M Nebuloni, E Sala, P Cinque, S Bonetto, A Isella, L Ottoni, A Crociati, G Costanzi
ABSTRACT
We evaluated the frequency and histopathological features of concomitant infections of the central nervous system (CNS) with cytomegalovirus (CMV) and herpes simplex viruses type 1 or 2 (HSV1/2) in a large series of patients who had died from AIDS. Eighty-two autopsy cases with a histological diagnosis of CMV necrotizing encephalitis were examined retrospectively. CMV and HSV1/2 were detected by immunohistochemistry (IHC) with poly- and monoclonal antibodies and by nested polymerase chain reaction (PCR) for HSV 1 and 2 on DNA extracted from paraffin blocks. PCR for a beta-globin genomic sequence was performed in all IHC-positive cases to verify the integrity of extracted DNA. Concomitant CMV/HSV infections were demonstrated by IHC in 13 cases (16%); using monoclonal antibodies, HSV1 was found in 9 cases and HSV2 in 4 cases. In half of the cases, HSV1- or HSV2-positive cells represented more than 25% of immunopositive CMV cells. In all 13 cases, double immunochemical staining showed cells containing both CMV and HSV antigens. PCR for HSV1 and 2 was positive in only 7 of 13 cases (5 HSV1 and 2 HSV2). In the remaining 6 negative cases PCR for beta-globin was also repeatedly negative. HSV1 or 2 infection can be demonstrated by IHC in a significant proportion of AIDS cases with necrotizing CMV encephalitis. Nested PCR for HSV1 and 2 on DNA extracted from formalin-fixed and paraffin-embedded autopsy tissues was positive in only slighty above 50% of IHC-positive cases.