MilliporeSigma
  • Immunohistochemistry identifies carriers of mismatch repair gene defects causing hereditary nonpolyposis colorectal cancer.
Immunohistochemistry identifies carriers of mismatch repair gene defects causing hereditary nonpolyposis colorectal cancer.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2005-07-22)
Astrid T Stormorken, Inger Marie Bowitz-Lothe, Tove Norèn, Elin Kure, Steinar Aase, Juul Wijnen, Jaran Apold, Ketil Heimdal, Pål Møller
ABSTRACT
Hereditary nonpolyposis colorectal cancer (HNPCC) may be caused by mutations in mismatch repair (MMR) genes. The aim of this study was to validate immunohistochemistry and family history as prescreening tools to predict germline mutations in MLH1, MSH2, and MSH6. Pedigrees from 250 families were extended, cancer diagnoses were verified, and families were classified according to the Amsterdam and the Bethesda criteria. Tumor specimens were examined with immunohistochemistry for the presence of MLH1, MSH2, and MSH6 proteins. Mutation analyses were performed in blood samples from the same patients. Blood samples from affected index persons in 181 families and tumor specimens from 127 of the affected index persons were obtained. Thirty tumors lacked one or more gene products. Sensitivity of immunohistochemistry to detect mutation carriers was 100%, specificity was 82%, and positive predictive value was 85%. Sensitivities, specificities, and positive predictive values for the Amsterdam criteria were 82%, 8%, and 45%, respectively, and for the Bethesda criteria were 100%, 0%, and 48%, respectively. Distribution of mutations was MLH1 = 4, MSH2 = 11, and MSH6 = 4. Wide clinical criteria to select HNPCC kindreds, followed by immunohistochemistry of tumor material from one affected person in each family, had high sensitivity and specificity to predict MMR mutations.