MilliporeSigma
  • A system for genome-wide histone variant dynamics in ES cells reveals dynamic MacroH2A2 replacement at promoters.
A system for genome-wide histone variant dynamics in ES cells reveals dynamic MacroH2A2 replacement at promoters.
PLoS genetics (2014-08-08)
Ozlem Yildirim, Jui-Hung Hung, Ryan J Cedeno, Zhiping Weng, Christopher J Lengner, Oliver J Rando
ABSTRACT
Dynamic exchange of a subset of nucleosomes in vivo plays important roles in epigenetic inheritance of chromatin states, chromatin insulator function, chromosome folding, and the maintenance of the pluripotent state of embryonic stem cells. Here, we extend a pulse-chase strategy for carrying out genome-wide measurements of histone dynamics to several histone variants in murine embryonic stem cells and somatic tissues, recapitulating expected characteristics of the well characterized H3.3 histone variant. We extended this system to the less-studied MacroH2A2 variant, commonly described as a "repressive" histone variant whose accumulation in chromatin is thought to fix the epigenetic state of differentiated cells. Unexpectedly, we found that while large intergenic blocks of MacroH2A2 were stably associated with the genome, promoter-associated peaks of MacroH2A2 exhibited relatively rapid exchange dynamics in ES cells, particularly at highly-transcribed genes. Upon differentiation to embryonic fibroblasts, MacroH2A2 was gained primarily in additional long, stably associated blocks across gene-poor regions, while overall turnover at promoters was greatly dampened. Our results reveal unanticipated dynamic behavior of the MacroH2A2 variant in pluripotent cells, and provide a resource for future studies of tissue-specific histone dynamics in vivo.
Product Description
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Doxycycline hyclate
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard