MilliporeSigma
Sublocalization of the human protein C gene on chromosome 2q13-q14.
Human genetics (1989-01-01)
P Patracchini, V Aiello, P Palazzi, E Calzolari, F Bernardi
ABSTRACT
The localization of human protein C gene on chromosome 2 was investigated by in situ hybridization using a partial cDNA for protein C. Silver-grain analysis indicates that the protein C gene is located on 2q13-q14.