MilliporeSigma
  • Prostate-specific membrane antigen as a marker of pancreatic cancer cells.
Prostate-specific membrane antigen as a marker of pancreatic cancer cells.
Medical oncology (Northwood, London, England) (2014-01-31)
He Ren, Huan Zhang, Xiuchao Wang, Junxiu Liu, Zhanna Yuan, Jihui Hao
ABSTRACT
The aim of this study was to identify the expression of prostate-specific membrane antigen (PSMA) and analyze the correlation between PSMA with clinical characteristics in patients with pancreatic cancer. The expression of PSMA protein and mRNA was detected by immunohistochemistry and real-time quantitative polymerase chain reaction in pancreatic cancer tissues, pancreatic intraepithelial neoplasia or normal pancreatic tissues, respectively. And clinical characteristics and prognosis of patients were investigated. PSMA was expressed in pancreatic cancer cells, both in protein and mRNA levels. Moreover, the PSMA levels were associated with the prognosis of patients with pancreatic ductal adenocarcinoma. The overall survival time of pancreatic cancer patients with high expression of PSMA was significantly shorter than that of the low ones. Moreover, the PSMA levels were correlated with clinicopathological features including the histological grade and pathological tumor-node-metastasis stage. PSMA is involved in the carcinogenesis of pancreatic cancer, and it might serve as a potential therapeutic target for pancreatic cancer.