MilliporeSigma
  • Biochemical and cellular evidence demonstrating AKT-1 as a binding partner for resveratrol targeting protein NQO2.
Biochemical and cellular evidence demonstrating AKT-1 as a binding partner for resveratrol targeting protein NQO2.
PloS one (2014-06-27)
Tze-chen Hsieh, Chia-Yi Lin, Dylan John Bennett, Erxi Wu, Joseph M Wu
ABSTRACT
AKT plays an important role in the control of cell proliferation and survival. Aberrant activation of AKT frequently occurs in human cancers making it an attractive drug targets and leading to the synthesis of numerous AKT inhibitors as therapeutic candidates. Less is known regarding proteins that control AKT. We recently reported that quinone reductase 2 (NQO2) inhibited AKT activity, by unknown mechanisms. In this study, molecular modeling was used to query interaction between NQO2 and AKT. We found that pleckstrin homology (PH) and kinase domains of AKT bind to chains A and B of NQO2. Pull-down and deletion assays revealed that PH domain of AKT is essential for interaction with NQO2. Modeling analysis further revealed that kinase domain of AKT binds NQO2 in the vicinity of asparagine 161 located in the resveratrol-binding domain of NQO2. In studies to test whether exposure to resveratrol potentiates or diminishes AKT binding to NQO2, we showed that pre-binding by resveratrol in wild type but not histidine-161 (N161H) mutant NQO2 significantly affected this interaction. To obtain information on interplay between resveratrol and AKT, resveratrol affinity chromatography was performed. AKT binds with high affinity to the column suggesting that it is a target of resveratrol. The half-life of AKT mRNA decreased from ∼4 h in control cells to ∼1 h in NQO2-knockdown cells. The inhibition of AKT by resveratrol was attenuated in NQO2-expressing relative to NQO2-knockdown cells. Both NQO2 and AKT are targets of resveratrol; NQO2:AKT interaction is a novel physiological regulator of AKT activation/function.
Product Description
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Akt1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Resveratrol, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%