MilliporeSigma
  • Exploring the anti-apoptotic role of HAX-1 versus BCL-XL in cytokine-dependent bone marrow-derived cells from mice.
Exploring the anti-apoptotic role of HAX-1 versus BCL-XL in cytokine-dependent bone marrow-derived cells from mice.
FEBS letters (2014-06-10)
Alicja Trebinska, Kari Högstrand, Alf Grandien, Ewa A Grzybowska, Bengt Fadeel
ABSTRACT
HS-1-associated protein X-1 (HAX-1) is a multi-functional protein that has been implicated in the regulation of apoptosis, cell motility and calcium homeostasis. In the present study, we set out to assess the postulated functional resemblance of HAX-1 to the BCL-2 family of anti-apoptotic proteins using non-transformed, cytokine-dependent murine bone marrow cells as a model system. BCL-X(L), but not HAX-1 protected against cytokine withdrawal-induced apoptosis while HAX-1 and BCL-X(L) significantly reduced thapsigargin-triggered (calcium-dependent) apoptosis. The data argue in favor of cell type- and stimulus-specific roles of HAX-1 in regulation of cell survival.
Product Description
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
L-Glutamine
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Orbifloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Orbifloxacin, VETRANAL®, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O