MilliporeSigma
  • Genes and gene networks involved in sodium fluoride-elicited cell death accompanying endoplasmic reticulum stress in oral epithelial cells.
Genes and gene networks involved in sodium fluoride-elicited cell death accompanying endoplasmic reticulum stress in oral epithelial cells.
International journal of molecular sciences (2014-05-24)
Yoshiaki Tabuchi, Tatsuya Yunoki, Nobuhiko Hoshi, Nobuo Suzuki, Takashi Kondo
ABSTRACT
Here, to understand the molecular mechanisms underlying cell death induced by sodium fluoride (NaF), we analyzed gene expression patterns in rat oral epithelial ROE2 cells exposed to NaF using global-scale microarrays and bioinformatics tools. A relatively high concentration of NaF (2 mM) induced cell death concomitant with decreases in mitochondrial membrane potential, chromatin condensation and caspase-3 activation. Using 980 probe sets, we identified 432 up-regulated and 548 down-regulated genes, that were differentially expressed by >2.5-fold in the cells treated with 2 mM of NaF and categorized them into 4 groups by K-means clustering. Ingenuity® pathway analysis revealed several gene networks from gene clusters. The gene networks Up-I and Up-II included many up-regulated genes that were mainly associated with the biological function of induction or prevention of cell death, respectively, such as Atf3, Ddit3 and Fos (for Up-I) and Atf4 and Hspa5 (for Up-II). Interestingly, knockdown of Ddit3 and Hspa5 significantly increased and decreased the number of viable cells, respectively. Moreover, several endoplasmic reticulum (ER) stress-related genes including, Ddit3, Atf4 and Hapa5, were observed in these gene networks. These findings will provide further insight into the molecular mechanisms of NaF-induced cell death accompanying ER stress in oral epithelial cells.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phenol Red, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium selenite, SAJ first grade, ≥90.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium selenite, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture