MilliporeSigma
  • Total synthesis and biological evaluation of the natural product (-)-cyclonerodiol, a new inhibitor of IL-4 signaling.
Total synthesis and biological evaluation of the natural product (-)-cyclonerodiol, a new inhibitor of IL-4 signaling.
Organic & biomolecular chemistry (2014-10-29)
Jens Langhanki, Kristina Rudolph, Gerhard Erkel, Till Opatz
ABSTRACT
In a screening program of natural compounds from fungi, the known cyclopentanoid sesquiterpene (-)-cyclonerodiol was identified as a specific inhibitor of the IL-4 induced STAT6 signaling pathway (IC50 = 9.7 μM) which is required for the differentiation of naive CD4 T cells to T helper type 2 (Th2) lymphocytes. As many allergic conditions, including allergic asthma and atopic diseases, are driven by an excessive Th2 response, STAT6 is a promising target for the development of new therapeutics. The compound was synthesized in six steps from (-)-linalool using an epoxide radical cyclization as the key step.