MilliporeSigma
  • Impact of conditional deletion of the pro-apoptotic BCL-2 family member BIM in mice.
Impact of conditional deletion of the pro-apoptotic BCL-2 family member BIM in mice.
Cell death & disease (2014-10-10)
M J Herold, R Stuchbery, D Mérino, T Willson, A Strasser, D Hildeman, P Bouillet
ABSTRACT
The pro-apoptotic BH3-only BCL-2 family member BIM is a critical determinant of hematopoietic cell development and homeostasis. It has been argued that the striking hematopoietic abnormalities of BIM-deficient mice (accumulation of lymphocytes and granulocytes) may be the result of the loss of the protein throughout the whole animal rather than a consequence intrinsic to the loss of BIM in hematopoietic cells. To address this issue and allow the deletion of BIM in specific cell types in future studies, we have developed a mouse strain with a conditional Bim allele as well as a new Cre transgenic strain, Vav-CreER, in which the tamoxifen-inducible CreER recombinase (fusion protein) is predominantly expressed in the hematopoietic system. We show that acute loss of BIM in the adult mouse rapidly results in the hematopoietic phenotypes previously observed in mice lacking BIM in all tissues. This includes changes in thymocyte subpopulations, increased white blood cell counts and resistance of lymphocytes to BIM-dependent apoptotic stimuli, such as cytokine deprivation. We have validated this novel conditional Bim knockout mouse model using established and newly developed CreER strains (Rosa26-CreER and Vav-CreER) and will make these exciting new tools for studies on cell death and cancer available.
Product Description
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Tamoxifen, ≥99%