- Phosphodiesterase 3A rs7134375 single nucleotide polymorphism and serum lipid levels.
Phosphodiesterase 3A rs7134375 single nucleotide polymorphism and serum lipid levels.
The association between the phosphodiesterase 3A (PDE3A) rs7134375 single nucleotide polymorphism (SNP) and serum lipid levels are not well understood in the general population. The present study was performed in order to detect the association between the rs7134375 SNP and serum lipid levels in the Guangxi Mulao and Han populations. The genotypes of the PDE3A rs7134375 SNP in 761 subjects of the Mulao population and 774 subjects of the Han Chinese population were determined by polymerase chain reaction and restriction fragment length polymorphism combined with gel electrophoresis, and then confirmed by direct sequencing. It was observed that serum low-density lipoprotein cholesterol and apolipoprotein B levels were higher in the Mulao population than in the Han population (P<0.05 for each). The frequencies of the C and A alleles were 72.14 and 27.86% in the Mulao population, and 78.55 and 21.45% in the Han population (P<0.01), respectively. The frequencies of the CC, CA and AA genotypes were 52.04, 40.21 and 7.75% in the Mulao population, and 61.50, 34.11 and 4.39% in the Han population (P<0.01), respectively. The frequencies of the C and A alleles were 74.89 and 25.11% in Mulao females, and 68.08 and 31.92% in Mulao males (P<0.01), respectively. The serum triglyceride (TG) levels were different among the genotypes in the Mulao population; however, not in the Han population (P<0.01), and the A allele carriers exhibited lower TG levels than the A allele noncarriers. The serum lipid parameters were also correlated with several environmental factors in the two ethnic groups (P<0.05-0.001). It was concluded that the genotypic and allelic frequencies of the rs7134375 SNP are different between the Mulao and Han populations. In addition, the PDE3A rs7134375 SNP is associated with serum TG levels in the Mulao population, however, not in the Han population.