MilliporeSigma
  • Downregulation of TBXAS1 in an iron-induced malignant mesothelioma model.
Downregulation of TBXAS1 in an iron-induced malignant mesothelioma model.
Cancer science (2015-07-28)
Daisuke Minami, Nagio Takigawa, Yuka Kato, Kenichiro Kudo, Hideko Isozaki, Shinsuke Hashida, Daijiro Harada, Nobuaki Ochi, Masanori Fujii, Toshio Kubo, Kadoaki Ohashi, Akiko Sato, Takehiro Tanaka, Katsuyuki Hotta, Masahiro Tabata, Shinichi Toyooka, Mitsune Tanimoto, Katsuyuki Kiura
ABSTRACT
Malignant mesothelioma is an aggressive and therapy-resistant neoplasm arising from mesothelial cells. Evidence suggests that the major pathology associated with asbestos-induced mesothelioma is local iron overload. In the present study, we induced iron-induced mesothelioma in rats based on previous reports. Ten Wistar rats were given ferric saccharate and nitrilotriacetate i.p. for 5 days a week. Five of the ten rats exhibited widespread mesotheliomas in the peritoneum and tunica vaginalis. The tumor cells showed positive immunostaining for calretinin, wilms tumor-1, podoplanin and the oxidative DNA marker 8-hydroxy-2'-deoxyguanosine. In three of the five rats with mesothelioma, array-based comparative genomic hybridization analysis identified a common chromosomal deletion mapped to the chromosomal 4q31 locus, which encompasses the TBXAS1 gene. Downregulation of the TBXAS1 gene was confirmed using quantitative PCR. TBXAS1 gene expression was also reduced in three of four human malignant pleural mesothelioma cell lines compared with normal bronchial epithelial cells. Immunohistochemistry revealed that TBXAS1 expression was weakly positive and positive in five and three out of eight human malignant mesothelioma samples, respectively. In conclusion, TBXAS1 gene expression was downregulated in rats with iron-induced mesothelioma. The relationship between iron overload and TBXAS1 downregulation should be pursued further.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%