MilliporeSigma
  • Expression, purification and preliminary crystallographic studies of human glutamate oxaloacetate transaminase 1 (GOT1).
Expression, purification and preliminary crystallographic studies of human glutamate oxaloacetate transaminase 1 (GOT1).
Protein expression and purification (2015-05-25)
Xiuping Jiang, Haiyang Chang, Yong Zhou
ABSTRACT
Glutamate oxaloacetate transaminase (GOT) catalyzes the reversible reaction of l-aspartate and α-ketoglutarate into oxaloacetate and L-glutamate and plays a key role in carbon and nitrogen metabolism in all organisms. In human tissues, GOTs are pyridoxal 5'-phosphate-dependent (PLP) enzymes which exist in cytoplasm and mitochondrial forms, GOT1 and GOT2, respectively. GOT1 expression correlates with the growth of several tumors because cancer cells can utilize the amino acid glutamine to fuel anabolic processes, and therefore, GOT1 represents a new therapeutic target in cancer. In this work, human GOT1 was expressed in Escherichia coli periplasmic space, and purified by a combination of His-tag immobilized metal-ion affinity chromatography and anion exchange chromatography. Optimal activity of the enzyme occurred at a temperature of 37 °C and a pH of 7.5. Cations such as Na(+), K(+) and Mg(2+) slightly inhibited the activity of recombinant human GOT1, while Zn(2+), Mn(2+), Cu(2+), Ni(2+), Co(2+) and Ca(2+) had stronger inhibitory effects. Crystals of human GOT1 were grown using the hanging-drop vapor diffusion method at 4°C with 0.1M Bis-Tris pH 6.0% and 21% (w/v) PEG 3350. The crystals diffracted to 2.99Å resolution and belonged to space group P43212 with the unit cell parameters a = b = 93.4, c = 107.4Å, α = β = γ = 90°.
Product Description
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O