MilliporeSigma
  • Heat shock protein 90 associates with Toll-like receptors 7/9 and mediates self-nucleic acid recognition in SLE.
Heat shock protein 90 associates with Toll-like receptors 7/9 and mediates self-nucleic acid recognition in SLE.
European journal of immunology (2015-04-15)
Keita Saito, Kazuharu Kukita, Goro Kutomi, Koichi Okuya, Hiroko Asanuma, Tetsuya Tabeya, Yasuka Naishiro, Motohisa Yamamoto, Hiroki Takahashi, Toshihiko Torigoe, Akira Nakai, Yasuhisa Shinomura, Koichi Hirata, Noriyuki Sato, Yasuaki Tamura
ABSTRACT
Systemic lupus erythematosus (SLE) is a prototype systemic autoimmune disease, and disease activity is associated with serum IFN-α level. Plasmacytoid dendritic cells (pDCs) sense microbial as well as self-nucleic acids by TLRs 7 and 9 and produce a large amount of IFN-α. Here, we show that heat shock protein 90 (Hsp90) associates with and delivers TLR7/9 from the ER to early endosomes for ligand recognition. Inhibition of Hsp90 by various approaches including the use of Hsp90 inhibitor, a geldanamycin derivative, suppressed the Hsp90 association with TLR7/9, which resulted in inhibition of IFN-α production, leading to improvement of SLE symptoms in mice. Notably, we observed that serum Hsp90 is clearly increased in patients with active SLE compared with that in patients with inactive disease. Furthermore, we demonstrated that serum Hsp90 detected in SLE patients binds to self-DNA and/or anti-DNA Ab, thus leading to stimulation of pDCs to produce IFN-α. Our data demonstrate that Hsp90 plays a crucial role in the pathogenesis of SLE and that an Hsp90 inhibitor will therefore provide a new therapeutic approach to SLE and other nucleic acid-related autoimmune diseases.
Sigma-Aldrich
DAPI, for nucleic acid staining
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF7
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O