MilliporeSigma
  • Bcl-2 phosphorylation is required for inhibition of oxidative stress-induced lysosomal leak and ensuing apoptosis.
Bcl-2 phosphorylation is required for inhibition of oxidative stress-induced lysosomal leak and ensuing apoptosis.
FEBS letters (2001-12-26)
M Zhao, J W Eaton, U T Brunk
ABSTRACT
B-cell leukemia/lymphoma 2 (Bcl-2) blocks oxidant-induced apoptosis at least partly by stabilizing lysosomes. Here we report that phosphorylation of Bcl-2 may be required for these protective effects. J774 cells overexpressing wild-type Bcl-2 resist oxidant-induced lysosomal leak as well as apoptosis, and this protection is amplified by pretreatment with phorbol 12-myristate 13-acetate (which promotes protein kinase C (PKC)-dependent phosphorylation of Bcl-2). In contrast, cells overexpressing the Bcl-2 mutant S70A (which cannot be phosphorylated) are not protected in either circumstance. Transfection with Bcl-2(S70E), a constitutively active Bcl-2 mutant which does not require phosphorylation, is protective independent of PKC activation. In contrast, C(2)-ceramide, a putative protein phosphatase 2A activator, abolishes the protective effects of wild-type Bcl-2 overexpression but does not diminish protection afforded by Bcl-2(S70E). Additional results suggest that, perhaps as a consequence of lysosomal stabilization, Bcl-2 may prevent activation of phospholipase A2, an event potentially important in the ultimate initiation of apoptosis.