MilliporeSigma
  • IL-32, a novel proinflammatory cytokine in chronic obstructive pulmonary disease.
IL-32, a novel proinflammatory cytokine in chronic obstructive pulmonary disease.
American journal of respiratory and critical care medicine (2008-08-16)
Fiorella Calabrese, Simonetta Baraldo, Erica Bazzan, Francesca Lunardi, Federico Rea, Piero Maestrelli, Graziella Turato, Kim Lokar-Oliani, Alberto Papi, Renzo Zuin, Paolo Sfriso, Elisabetta Balestro, Charles A Dinarello, Marina Saetta
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is a chronic inflammatory disorder of the lung, yet the mechanisms that regulate this immune-inflammatory response are not fully understood. We investigated whether IL-32, a newly discovered cytokine, was related to markers of inflammation and clinical progression in COPD. Using immunohistochemistry, expression of IL-32 was examined in surgically resected specimens from 40 smokers with COPD (FEV(1) = 39 +/- 4% predicted), 11 smokers with normal lung function, and 9 nonsmoking control subjects. IL-32 was quantified in alveolar macrophages, alveolar walls, bronchioles, and arterioles, and confirmed by molecular analysis. The levels of IL-32 were correlated with the cellular infiltrates, markers of inflammation, and clinical data. Macrophage staining for IL-32 was increased in smokers with COPD compared with control smokers and nonsmokers (P = 0.0014 and P < 0.0001, respectively), and similar differences were observed in alveolar walls (P = 0.0004 and P = 0.0005) and bronchiolar epithelium (P = 0.004 and P = 0.0009). This increase was also detected at the mRNA level (P = 0.007 vs. control smokers and P = 0.029 vs. nonsmokers) and was mainly due to non-alpha isoforms. Moreover, IL-32 expression was positively correlated with tumor necrosis factor-alpha (P = 0.004, r(s)=0.70), CD8(+)cells (P = 0.02, r(s)=0.46), phospho p38MAPK (P < 0.01, r(s)=0.60) and negatively with FEV(1) values (P = 0.004, r(s)= -0.53). This is the first study to demonstrate increased expression of IL-32 in lung tissue of patients with COPD, where it was colocalized with tumor necrosis factor-alpha and correlated with the degree of airflow obstruction. These results suggest that IL-32 is implicated in the characteristic immune response of COPD, with a possible impact on disease progression.