- Membrane chromatographic immunoassay method for rapid quantitative analysis of specific serum antibodies.
Membrane chromatographic immunoassay method for rapid quantitative analysis of specific serum antibodies.
This paper discusses a membrane chromatographic immunoassay method for rapid detection and quantitative analysis of specific serum antibodies. A type of polyvinylidine fluoride (PVDF) microfiltration membrane was used in the method for its ability to reversibly and specifically bind IgG antibodies from antiserum samples by hydrophobic interaction. Using this form of selective antibody binding and enrichment an affinity membrane with antigen binding ability was obtained in-situ. This was done by passing a pulse of diluted antiserum sample through a stack of microporous PVDF membranes. The affinity membrane thus formed was challenged with a pulse of antigen solution and the amount of antigen bound was accurately determined using chromatographic methods. The antigen binding correlated well with the antibody loading on the membrane. This method is direct, rapid and accurate, does not involve any chemical reaction, and uses very few reagents. Moreover, the same membrane could be repeatedly used for sequential immunoassays on account of the reversible nature of the antibody binding. Proof of concept of this method is provided using human hemoglobin as model antigen and rabbit antiserum against human hemoglobin as the antibody source.