MilliporeSigma
  • Phosphorylation of Williams syndrome transcription factor by MAPK induces a switching between two distinct chromatin remodeling complexes.
Phosphorylation of Williams syndrome transcription factor by MAPK induces a switching between two distinct chromatin remodeling complexes.
The Journal of biological chemistry (2009-09-25)
Hiroyuki Oya, Atsushi Yokoyama, Ikuko Yamaoka, Ryoji Fujiki, Masayoshi Yonezawa, Min-Young Youn, Ichiro Takada, Shigeaki Kato, Hirochika Kitagawa
ABSTRACT
Changes in the environment of a cell precipitate extracellular signals and sequential cascades of protein modification and elicit nuclear transcriptional responses. However, the functional links between intracellular signaling-dependent gene regulation and epigenetic regulation by chromatin-modifying proteins within the nucleus are largely unknown. Here, we describe novel epigenetic regulation by MAPK cascades that modulate formation of an ATP-dependent chromatin remodeling complex, WINAC (WSTF Including Nucleosome Assembly Complex), an SWI/SNF-type complex containing Williams syndrome transcription factor (WSTF). WSTF, a specific component of two chromatin remodeling complexes (SWI/SNF-type WINAC and ISWI-type WICH), was phosphorylated by the stimulation of MAPK cascades in vitro and in vivo. Ser-158 residue in the WAC (WSTF/Acf1/cbpq46) domain, located close to the N terminus of WSTF, was identified as a major phosphorylation target. Using biochemical analysis of a WSTF mutant (WSTF-S158A) stably expressing cell line, the phosphorylation of this residue (Ser-158) was found to be essential for maintaining the association between WSTF and core BAF complex components, thereby maintaining the ATPase activity of WINAC. WINAC-dependent transcriptional regulation of vitamin D receptor was consequently impaired by this WSTF mutation, but the recovery from DNA damage mediated by WICH was not impaired. Our results suggest that WSTF serves as a nuclear sensor of the extracellular signals to fine-tune the chromatin remodeling activity of WINAC. WINAC mediates a previously unknown MAPK-dependent step in epigenetic regulation, and this MAPK-dependent switching mechanism between the two functionally distinct WSTF-containing complexes might underlie the diverse functions of WSTF in various nuclear events.
Sigma-Aldrich
Akt Inhibitor X, The Akt Inhibitor X, also referenced under CAS 925681-41-0, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-Myc Tag Antibody, Upstate®, from rabbit
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution