MilliporeSigma
Pericellular-acting proteases in human first trimester decidua.
Molecular human reproduction (2008-01-08)
Margreet Plaisier, Pieter Koolwijk, Florian Willems, Frans M Helmerhorst, Victor W M van Hinsbergh
ABSTRACT
Proteolysis is essential for decidual development during embryonic implantation, but little is known regarding the expression and functions of membrane-type matrix metalloproteinases (MT-MMPs) and urokinase-type plasminogen activator (uPA) and its receptor uPAR in decidua. Therefore, their protein and mRNA levels were analysed in three first trimester decidual tissues, decidual secretory endometrium (DSE), decidua parietalis (DP) and basalis (DB). Decidua was obtained during first trimester pregnancy termination. uPA, uPAR, and MT1/2/3/5-MMP expression were studied by RT-PCR and immunohistochemistry, and CD56-positive uNK cells and CD68-positive macrophages were quantified in serial sections. The mRNAs and antigens of all proteases and uPAR were detectable in the decidual tissues and extravillous trophoblasts (EVT). mRNA levels of all proteases and uPAR, except MT5-MMP, were elevated in both DB and DP compared to DSE, being significant for MT1-MMP and uPAR in DP. MT2- and MT3-MMP mRNAs in DB were 24- and 10-fold higher than in DSE, and 19- and 7-fold increased compared to DP. At the protein level uPA and uPAR were particularly elevated in DB, while pro-angiogenic MT1- and MT3-MMPs were elevated in both DB and DP compared to DSE. MT2-MMP was prominently present in all conditions. The number of uNK cells was increased in DB and DP versus DSE, while a comparable increase in macrophages did not reach statistical significance. These data are consistent with a differential regulation of pericellular proteases in decidua by pregnancy-induced hormones, immune cells and EVT.