MilliporeSigma
  • IgE cross reactivity between reindeer and bovine milk beta-lactoglobulins in cow's milk allergic patients.
IgE cross reactivity between reindeer and bovine milk beta-lactoglobulins in cow's milk allergic patients.
Journal of investigational allergology & clinical immunology (2006-10-17)
T J Suutari, K H Valkonen, T J Karttunen, B M Ehn, B Ekstrand, U Bengtsson, V Virtanen, M Nieminen, J Kokkonen
ABSTRACT
Allergic reactions to cow's milk are common in small children. One of the main protein allergens found in cow's milk is beta-lactoglobulin (beta-Lg). Reindeer and bovine milk both contain related beta-Lg proteins, but the allergenicity of reindeer beta-Lg has not previously been studied. The purpose of this study was to analyze the immunological cross-reactivity of IgE antibodies from children with cow's milk allergy to reindeer and bovine beta-Lg. Sera from 17 children and a serum pool of 4 patients with elevated cow's milk-specific IgE were investigated. Beta-Lg from bovine and reindeer milk was isolated in native form and an enzyme-linked immunosorbent inhibition assay was developed. Bovine beta-Lg was used as a capturing antigen and the inhibiting effects of reindeer and bovine beta-Lg on the IgE binding were measured. Cross-reactivity patterns of bovine milk beta-Lg specific IgE to reindeer beta-Lg varied among patients. In general, reindeer beta-Lg showed significantly lower inhibition (mean 43%) of IgE binding to the capturing antigen than did bovine beta-Lg (mean 89%). In some patients, even high concentrations of reindeer beta-Lg only partly eliminated the IgE binding to bovine beta-Lg. The partial cross-reactivity of human anti-bovine IgE with reindeer beta-Lg suggests that it lacks important bovine epitopes and those that are recognized are only weakly bound.