MilliporeSigma
  • Engineered Context-Sensitive Agonism: Tissue-Selective Drug Signaling through a G Protein-Coupled Receptor.
Engineered Context-Sensitive Agonism: Tissue-Selective Drug Signaling through a G Protein-Coupled Receptor.
The Journal of pharmacology and experimental therapeutics (2017-01-14)
Wiebke K Seemann, Daniela Wenzel, Ramona Schrage, Justine Etscheid, Theresa Bödefeld, Anna Bartol, Mareille Warnken, Philipp Sasse, Jessica Klöckner, Ulrike Holzgrabe, Marco DeAmici, Eberhard Schlicker, Kurt Racké, Evi Kostenis, Rainer Meyer, Bernd K Fleischmann, Klaus Mohr
ABSTRACT
Drug discovery strives for selective ligands to achieve targeted modulation of tissue function. Here we introduce engineered context-sensitive agonism as a postreceptor mechanism for tissue-selective drug action through a G protein-coupled receptor. Acetylcholine M
Sigma-Aldrich
Oxotremorine M, solid