MilliporeSigma
  • The mechanism of sirtuin 2-mediated exacerbation of alpha-synuclein toxicity in models of Parkinson disease.
The mechanism of sirtuin 2-mediated exacerbation of alpha-synuclein toxicity in models of Parkinson disease.
PLoS biology (2017-03-04)
Rita Machado de Oliveira, Hugo Vicente Miranda, Laetitia Francelle, Raquel Pinho, Éva M Szegö, Renato Martinho, Francesca Munari, Diana F Lázaro, Sébastien Moniot, Patrícia Guerreiro, Luis Fonseca-Ornelas, Zrinka Marijanovic, Pedro Antas, Ellen Gerhardt, Francisco Javier Enguita, Bruno Fauvet, Deborah Penque, Teresa Faria Pais, Qiang Tong, Stefan Becker, Sebastian Kügler, Hilal Ahmed Lashuel, Clemens Steegborn, Markus Zweckstetter, Tiago Fleming Outeiro
ABSTRACT
Sirtuin genes have been associated with aging and are known to affect multiple cellular pathways. Sirtuin 2 was previously shown to modulate proteotoxicity associated with age-associated neurodegenerative disorders such as Alzheimer and Parkinson disease (PD). However, the precise molecular mechanisms involved remain unclear. Here, we provide mechanistic insight into the interplay between sirtuin 2 and α-synuclein, the major component of the pathognomonic protein inclusions in PD and other synucleinopathies. We found that α-synuclein is acetylated on lysines 6 and 10 and that these residues are deacetylated by sirtuin 2. Genetic manipulation of sirtuin 2 levels in vitro and in vivo modulates the levels of α-synuclein acetylation, its aggregation, and autophagy. Strikingly, mutants blocking acetylation exacerbate α-synuclein toxicity in vivo, in the substantia nigra of rats. Our study identifies α-synuclein acetylation as a key regulatory mechanism governing α-synuclein aggregation and toxicity, demonstrating the potential therapeutic value of sirtuin 2 inhibition in synucleinopathies.
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT2
Sigma-Aldrich
Anti-Aggregated a-Synuclein Antibody, clone 5G4, clone 5G4, from mouse, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit