- Synthesis and Evaluation of the Novel Prostamide, 15-Deoxy, Δ
Synthesis and Evaluation of the Novel Prostamide, 15-Deoxy, Δ
Molecular cancer therapeutics (2017-03-16)
Daniel A Ladin, Eman Soliman, Rene Escobedo, Timothy L Fitzgerald, Li V Yang, Colin Burns, Rukiyah Van Dross
PMID28292936
ABSTRACT
15-deoxy, Δ