MilliporeSigma
  • Quercetin promotes motor and sensory function recovery following sciatic nerve-crush injury in C57BL/6J mice.
Quercetin promotes motor and sensory function recovery following sciatic nerve-crush injury in C57BL/6J mice.
The Journal of nutritional biochemistry (2017-05-02)
Ming-Ming Chen, Jing Qin, Shu-Jian Chen, Le-Meng Yao, Lu-Yong Zhang, Zhi-Qi Yin, Hong Liao
ABSTRACT
Injuries and diseases that occur in the nervous system are common and have few effective treatments. Previous studies have shown that quercetin has a therapeutic effect on nervous system injuries, but its potential effects on and mechanisms of action related to behavioral recovery and axonal regrowth have not been investigated. Here, we showed that quercetin administration promotes behavioral recovery following sciatic nerve-crush injury in mice. Long-term evaluation showed that mice administered 20 mg·kg