Merck
1.17328
Millipore
MAS-100 Atmos® Microbial Compressed Gas Sampler
greener alternative
For Compressed Gas Sampling
Đăng nhậpđể Xem giá theo tổ chức & hợp đồng
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
Tài liệu quan trọng
Dịch vụ kỹ thuật
Bạn cần trợ giúp? Đội ngũ các nhà khoa học có kinh nghiệm của chúng tôi đang ở đây vì các bạn.
Hãy để chúng tôi hỗ trợ
Dịch vụ kỹ thuật
Bạn cần trợ giúp? Đội ngũ các nhà khoa học có kinh nghiệm của chúng tôi đang ở đây vì các bạn.
Hãy để chúng tôi hỗ trợ
material
polystyrene housing
reg. compliance
FDA
product line
MAS-100®
feature
portable
manufacturer/tradename
MAS-100 Atmos®
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with 90 mm Petri Dishes
greener alternative category
General description
The portable and battery-operated MAS-100 Atmos® is designed for failure-safe and accurate sampling of pre-selectable compressed gases.
Like all MAS-100® microbial air samplers, the equipment principle is based on sieve impaction. The impaction of microorganisms takes place under pressure to prevent any harm to microorganisms caused by fast dynamic decompression.
A specific amount of compressed gas gets aspirated through a perforated lid (300*0.6mm) which captures the available microbes onto the surface of an agar plate (90-100mm pertridish) with an impaction speed of 19.7m/s.
The state-of-art design provides a nominal d50 value of 1.1 μm at a default flow rate of 100 liter/min (LPM).
An optional perforated lid with 300x0.47mm holes can be used for pre-selected flow rates of 50 LPM to achieve the same d50.
The system measures the current pressure, indicated either in bar abs. or Psi (1.2 to 7 bar abs. = 17.4 to 101.5 Psi), and regulates the flow rate based on a mass-flow sensor for each pre-selected gas type to 100 LPM. In case of low flow provided by the gas pipe, the instrument can automatically switch to 50 LPM. The decompression after sampling is performed automatically.
Data Integrity:
The MAS-100 Atmos® offers various operation modes for different workflow scenarios configurable via browser-based user interface:
  • Freely accessible configuration by any operator (out of the box)
  • 21 CFR Part 11 compliant management.
  • Manage connected bar code reader.
  • 2 D bar code including all sample data supports wireless transfer into EM software systems
A full audit trail including e-signatures as well as sample reports and error logs are available for download.
Individual configurable hardware keys combined with a PIN code allow 21 CFR Part 11 compliant login including defined access rights the selected user role as well as traceability.
The touch screen is used for operation and to display important information, such as current pressure (units selectable in bar abs. or Psi), battery status, date and time, sampling status and result data as a 2D bar code. A graphical user guide can be selected for easy instructions of workflow steps.
The MAS-100 Atmos® is powered by a Lithium-ion rechargeable battery that assures long battery life without routine discharging.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS score card.
Application
MAS-100 Atmos® Microbial Compressed Gas Sampler is used to sample pre-defined compressed gases like argon, carbon dioxide, compressed air, and nitrogen from pharmaceutical, cosmetic, food & beverage industry.
Features and Benefits
  • Easy to handle including graphical user guide on touch screen.
  • Fully automated flow regulation per gas type by a mass flow sensor and pressure sensor.
  • Fully automated decompression cycle.
  • Software fully integrated in instrument and manageable via various web-browsers with internet connection.
  • 21 CFR Part 11 compliant user management.
  • Individual hardware key for login.
  • Sample Data as barcode on touch screen for wireless integration into EM software programs using barcode readers.
  • Impaction under pressure to reduces the risk of false negative results.
Components
  • 1 x MAS-100 Atmos® microbial compressed gas sampler.
  • 1 x Perforated lid 300x0.6mm.
  • 1 x Dust Cover.
  • 1 x Pressure connection tube; 2m PTFE; including 3/8′′ BSP female connector.
  • 1 x Mini Trip-clamp (Ø 34 mm)
  • 1 x Silicone gasket.
  • 1 x Battery Charger including USB-C cable and country specific plug adapters.
  • Documentation: Quick-start guide, declaration of conformity and calibration certificates (MAS-100 Atmos and calibration tool)
Other Notes
Operation of the pump is easy, and the following steps can be followed:
1.Insert the appropriate agar plate and select sample parameter (gas type, volume)
2.Connect the instrument to the gas outlet (PTFE pressure connection tube is provided with a 3/8′′BSP female adapter)
3.Open the gas valve and start sampling.
4.After sampling close the valve and start decompression.
5.Remove the plate.
6.Incubate the plate and count the colonies.
7.Correct CFU number according Feller table.
8.Calculate CFU/m3 (1000 Liter).
Contamination or intoxication of the sampling environment can be prevented mounting the optional exhaust tubing.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Atmos is a registered trademark of MBV AG
Lớp lưu trữ
11 - Combustible Solids
Giấy chứng nhận phân tích (COA)
Tìm kiếm Giấy chứng nhận phân tích (COA) bằng cách nhập Số lô của sản phẩm. Có thể tìm thấy Số lô trên nhãn sản phẩm theo sau từ ‘Lô’.
Đã sở hữu sản phẩm này?
Tìm tài liệu về sản phẩm bạn mua gần đây trong Thư viện tài liệu.
Truy cập thư viện tài liệu
Đội ngũ nhà khoa học của chúng tôi có kinh nghiệm trong mọi lĩnh vực nghiên cứu bao gồm Khoa học đời sống, Khoa học vật liệu, Tổng hợp hóa học, Sắc ký, Phân tích và nhiều lĩnh vực khác.
Liên hệ với dịch vụ kỹ thuật