biological source
synthetic
Quality Level
form
liquid
species reactivity
human, rat, mouse
concentration
200 μg/mL
shipped in
wet ice
storage temp.
−20°C
1 of 4
This Item | SAB4501460 | ABC97 | MABC535 |
---|---|---|---|
biological source rabbit | biological source rabbit | biological source rabbit | biological source rat |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
species reactivity mouse | species reactivity rat, human, mouse | species reactivity human | species reactivity mouse, human |
Immunogen
Application
Linkage
Disclaimer
related product
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service