MABN2186
Anti-DPP6 Antibody, clone 7G1.1
clone 7G1.1, from mouse
Synonym(s):
Dipeptidyl aminopeptidase-like protein 6, DPPX, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, Dipeptidyl peptidase VI, DPP VI
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
7G1.1, monoclonal
species reactivity
rat, mouse, human
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... DPP6(1804)
General description
Specificity
Immunogen
Application
Neuroscience
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected DPP6 in human cerebral cortex, human cerebellum, rat cerebral cortex, and rat cerebellum tissue.
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected DPP6 in 10 µg of mouse brain tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service