Merck
Z124559
Sigma-Aldrich
Suba-Seal® septa
red rubber, Suba-Seal® 9, fits neck I.D., 8 mm
About This Item
Código UNSPSC:
41121800
eCl@ss:
32119211
NACRES:
NB.35
Assistência Técnica
Precisa de ajuda? Nossa equipe de cientistas experientes está à sua disposição.
Estamos aqui para ajudar
Assistência Técnica
Precisa de ajuda? Nossa equipe de cientistas experientes está à sua disposição.
Estamos aqui para ajudar
Materiais
red rubber
Características
fits neck I.D. 8 mm
fabricante/nome comercial
Suba-Seal® 9
Categorias relacionadas
Descrição geral
Well-known as the best quality rubber turn-over closure with serrated stopper, Suba-Seal brand is available exclusively from Sigma-Aldrich. Ideal for air and moisture-sensitive chemistry procedures. After piercing with non-coring needle, rubber closes puncture, providing air and moisture tight seal to protect contents of vessel or container from atmosphere.
This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suctions sealing point.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings.
Informações legais
Suba-Seal is a registered trademark of Merck KGaA, Darmstadt, Germany
Escolha uma das versões mais recentes:
Certificados de análise (COA)