Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

930849

Sigma-Aldrich

[1,3-Bis[2-(diphenylmethyl)-4,6-dimethylphenyl]-1H-imidazolyl-2-yl]Ni(Cp)Cl

Synonym(s):

Chloro(cyclopentadienyl){1,3-bis[2-(diphenylmethyl)-4,6-dimethylphenyl]1H-imidazolium}nickel(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H45ClN2Ni
CAS Number:
Molecular Weight:
768.05
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.21

form

powder

Quality Level

storage temp.

−20°C

SMILES string

CC1=CC(C)=CC(C(C2=CC=CC=C2)C3=CC=CC=C3)=C1N4C([Ni]Cl)N(C5=C(C(C6=CC=CC=C6)C7=CC=CC=C7)C=C(C)C=C5C)C=C4.c8cccc8

Application

[1,3-Bis[2-(diphenylmethyl)-4,6-dimethylphenyl]-1H-imidazolyl-2-yl]Ni(Cp)Cl is a Ni NHC catalyst for a variety of cross-coupling reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service