Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

935514

Sigma-Aldrich

Rhodium(III) sulfate

anhydrous, powder, ≥99.9% trace metals basis

Synonym(s):

Rhodium Sulfate, Rhodium trisulfate, Rhodium(III) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O12Rh2S3
CAS Number:
Molecular Weight:
494.00
UNSPSC Code:
12352302

grade

anhydrous

Quality Level

Assay

≥99.9% trace metals basis
40.2-43.01% rhodium (Rh) basis (gravimetric)

form

powder

impurities

≤1000 ppm trace metals

color

beige-brown to very dark brown

solubility

water: very soluble(lit.)

density

1.217 g/cm3

storage temp.

2-8°C

SMILES string

[Rh].O=S(=O)(O)O

InChI

1S/H2O4S.Rh/c1-5(2,3)4;/h(H2,1,2,3,4);

InChI key

NZVSOIFAVIORHD-UHFFFAOYSA-N

General description

Anhydrous rhodium(III)sulfate is a reddish yellow crystalline solid. At 400°C, rhodium metal and sulfuric acid are used to produce the anhydrous form.

Application

Rhodium(III)sulfate is a plating reagent and noble metal additive. It is used as a reducing agent in metal surface analysis and as an oxidation catalyst in organic reactions.Furthermore, because of its potential for usage in fuel cells, Rhodium(III)sulfate is added to Platinum electrocatalyst to form Pt/Rh alloy in the research of ethanol oxidation. It demonstrates the ability to break the C-C bond and promotes the breaking of the C-H bond.

Rhodium(III)sulfate is also used with Platinum to form Pt/Rh alloy for highly corrosion resistant coatings on a metal substrate stainless steel (SS304). The alloy of Pt/Rh has been also used study the MB dye degaradtion using photocatalysis method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service