Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

935808

Sigma-Aldrich

Multiphoton lithography bioink kit

synthetic hydrogel bioink

Synonym(s):

HYDROTECH INX X100

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

form

liquid

color

yellow-orange

viscosity

0.5-5 Pa.s

gel strength

(Youngs modulus 30-40 MPa)

Looking for similar products? Visit Product Comparison Guide

General description

This synthetic bioink for multiphoton lithography (MPL) based 3D bioprinting applications can be used to generate hydrogel microstructures and complex architectures for tissue engineering applications. It is biologically inert but can be coated to allow cell adhesion and proliferation. The synthetic bioink hydrogel kit is a ready-to-use formulation that can be processed via a MPL-based printer after a short preheating process. The resin can be processed at high scanning speeds (up to 600 mm/s) which is favorable for shorter fabrication times.

Application

  • 3D bioprinting
  • Tissue engineering
  • High resolution printing of complex and open geometries with mechanical robustness

Features and Benefits

  • Biocompatibility - expression biocompatibility with no toxic effect on living cells
  • Stability - Forms a biostable hydrogel that sustain 3D cellular structures making it suitable for long term applications
  • Proccessability - Easy processing into open and complex architectures with minimal deformation
  • Mechanical integrity - Very robust hydrogels suitable for stiff tissue engineering applications
  • Easy to handle - Provided as ready to print formulation
  • Reproducibility - Production under strict quality control to provide a material that delivers every time

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.9 °F

Flash Point(C)

-14.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service