Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

936138

Sigma-Aldrich

N1-([1,1′-Biphenyl]-2-yl)-N2-(2-phenylnaphthalen-1-yl)benzene-1,2-diamine

new
greener alternative

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H26N2
CAS Number:
Molecular Weight:
462.58
UNSPSC Code:
12352200

Quality Level

Assay

≥95%

form

powder or crystals

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

mp

151-155 °C

greener alternative category

storage temp.

−20°C

SMILES string

C1(NC2=C(C=CC=C3)C3=CC=C2C4=CC=CC=C4)=CC=CC=C1NC5=C(C6=CC=CC=C6)C=CC=C5

InChI key

XDBPZHWQLWXPJP-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

N1-([1,1′-biphenyl]-2-yl)-N2-(2-phenylnaphthalen-1-yl)benzene-1,2-diamine is an anionic diamine ligand for copper catalysis. The ligand has been used in:
  • The Cu-catalyzed amination of aryl bromides under mild conditions.
  • The Room-Temperature Copper-Catalyzed Etherification of Aryl Bromides.
  • The Copper-Catalyzed Amination of Aryl Chlorides under Mild Reaction Conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service