Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

938734

Sigma-Aldrich

EPhos Pd G6 Br

greener alternative

Synonym(s):

(EPhos)Pd(p-CF3C6H4)(Br)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H59BrF3OPPd
Molecular Weight:
866.22
UNSPSC Code:
12352100
NACRES:
NA.21

form

powder

Quality Level

reaction suitability

core: palladium
reaction type: Cross Couplings
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

faint yellow

functional group

phosphine

greener alternative category

SMILES string

CC(C)OC1=CC=CC(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=C1[P]([Pd](Br)C3=CC=C(C(F)(F)F)C=C3)(C4CCCCC4)C5CCCCC5

InChI

InChI=1S/C36H55OP.C7H4F3.BrH.Pd/c1-24(2)28-22-32(25(3)4)35(33(23-28)26(5)6)31-20-15-21-34(37-27(7)8)36(31)38(29-16-11-9-12-17-29)30-18-13-10-14-19-30;8-7(9,10)6-4-2-1-3-5-6;;/h15,20-27,29-30H,9-14,16-19H2,1-8H3;2-5H;1H;

InChI key

BUVPEGXQNXKJOG-UHFFFAOYSA-N

General description

(EPhos Pd G6 Br) is a G6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

The Buchwald G6 precatalysts are oxidative addition complexes (OACs), which exhibit the same advantages as the previous generations of Buchwald precatalysts: quantitative generation of L–Pd(0), thermal stability, air stability, moisture stability, ease of handling, and high efficiency.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service