Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

939609

Sigma-Aldrich

[SIPr-H]2Pd2Cl6

greener alternative

Synonym(s):

Bis-(1,3-bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium)-hexachloropalladate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H78Cl6N4Pd2
CAS Number:
Molecular Weight:
1208.78
UNSPSC Code:
12352100

form

powder or crystals

Quality Level

reaction suitability

reagent type: catalyst
core: palladium
reaction type: Buchwald-Hartwig Amination

reagent type: catalyst
core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: catalyst
core: palladium
reaction type: Cross Couplings

reagent type: catalyst
core: palladium
reaction type: Heck Reaction

reagent type: catalyst
core: palladium
reaction type: Hiyama Coupling

reagent type: catalyst
core: palladium
reaction type: Negishi Coupling

reagent type: catalyst
core: palladium
reaction type: Sonogashira Coupling

reagent type: catalyst
core: palladium
reaction type: Stille Coupling

reagent type: catalyst
core: palladium
reaction type: Suzuki-Miyaura Coupling

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

brown to orange

greener alternative category

SMILES string

CC(C(C=CC=C1C(C)C)=C1N2CC[N+](C3=C(C=CC=C3C(C)C)C(C)C)=C2)C.Cl[Pd]4(Cl)[ClH][Pd](Cl)(Cl)[ClH]4.CC(C(C=CC=C5C(C)C)=C5N6CC[N+](C7=C(C=CC=C7C(C)C)C(C)C)=C6)C

InChI

InChI=1S/2C27H39N2.4ClH.2Cl.2Pd/c2*1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;;;;;;;;/h2*9-14,17-21H,15-16H2,1-8H3;4*1H;;;;/q2*+1;;;;;;;2*+2/p-4

InChI key

WHCUDDZJJCQTRX-UHFFFAOYSA-J

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.
[SIPr-H]2Pd2Cl6 is an ionic tetrachloropalladate complex containing 2 equivalents of the cationic imadazolium ligand 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium.

Application

Upon treatmeant with a suitable base, a catalytically active Pd-NHC complex can be generated in-situ. Suitable for use in difficult Suzuki-Miyaura and Buchwald-Hartwig cross-couplings reactions of aryl chlorides, and trans-amidation cross-coupling reactions of esters and amides.

Features and Benefits

[SPr-H]2Pd2Cl6 offers similar or greater performance to other similar Pd-NHC precatalysts available while offering several major advatages:
  • The precatalyst avoids the use of ′throw away′ stabilizing ligands often used in similar Pd-NHC complexes.
  • The more sustainable synthesis of [SIPr-H]2Pd2Cl6 compared to similar Pd-NHC complexes makes the product a more sustainable choice.
  • A competitive price, with a higher Pd percentage than many other Pd-NHC catalysts.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service