Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

940542

Lithium germanium phosphorus sulfide

new

(LGPS) solid electrolyte, powder

Synonym(s):

Sulfide solid electrolyte

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
GeLi10P2S12
CAS Number:
Molecular Weight:
588.78
UNSPSC Code:
12352302

grade

battery grade

Quality Level

description

application(s): battery manufacturing

Assay

99.9%

form

powder

mol wt

588.69 g/mol

composition

Li10GeP2S12

density

2.0 g/mL

application(s)

battery manufacturing

SMILES string

[Li].[S].[Ge].S=P([S-])([S-])[S-]

General description

Lithium germanium phosphorus sulfide solid electrolyte powder is a ceramic electrolyte with high ionic conductivity. Li10GeP2S12 (LGPS) is a white powder that crystallizes in the P42/nmc space group with three-dimensional interconnecting channels of Li+ ions that allow for fast Li+ diffusion. It belongs to the class of sulfide electrolytes and should be handled in a glovebox because LGPS is sensitive to air and moisture.

Application

Li10GeP2S12 (LGPS) is a solid electrolyte powder used in all-solid-state lithium batteries. It has an extremely high ionic conductivity of 12 mS cm–1 which is even higher than those of organic liquid electrolytes currently used in practical lithium-ion batteries. Typically, scientists pelletize (cold-press) the powder and may optionally anneal the pellet at high temperature (550 C) to enhance the ionic conductivity. LGPS is unstable against Li metal and is often used as the catholyte.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service