Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

990445

Sigma-Aldrich

Lithium manganese iron phosphate (LMFP)

new

Electrode sheet, aluminum substrate, size 5 in. x 10 in.

Synonym(s):

LMFP tape, Lithium iron manganese phosphate cathode

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiMn(x)Fe(x-1)PO4
UNSPSC Code:
26111700

grade

battery grade

Quality Level

description

3.8V vs. Li/Li+

form

sheet

composition

Active material loading 8.9 mg/cm2 ± 5%, ≥85% by wt (excluding current collector)

film size

80 μm × 16 μm , Al current collector

particle size

2 μm (active material characteristic)

capacity

(Areal capacity 1.25 mAh/cm2)
(First discharge capacity ≥140 mAh/g)

application(s)

battery manufacturing

SMILES string

[Li].[Mn].[Fe].O=P([O-])([O-])[O-]

InChI key

ZWXLZABZHZXDGX-UHFFFAOYSA-K

General description

Our lithium manganese iron phosphate (LMFP) electrode sheet is a ready-to-use cathode designed for lithium-ion battery research. The LMFP cathode film is 80 µm thick, single-sided, and applied to a 16 µm thick aluminum foil current collector measuring 5 × 10 inches (127 mm × 254 mm). The composition consists of 88% lithium iron phosphate (LFP), 4% Poly(vinylidene fluoride) [PVDF], and 8% carbon black. The active material, olivine LiMnxFe1−xPO4, is a well-studied cathode material known for its excellent cycling stability.

Application

Our lithium manganese iron phosphate (LMFP) electrode serves as a cathode in lithium-ion battery research. It is cost-effective, environmentally friendly, and cobalt-free. Compared to lithium iron phosphate (LFP), LMFP boasts a nearly 20% higher energy density due to its higher nominal voltage (3.8 V for LMFP vs. 3.2 V for LFP). Moreover, LMFP offers high specific capacity (>150 mAh/g) and a longer cycle life compared to most other lithium-ion chemistries. LMFP cells experience a slower rate of capacity loss and greater calendar-life than other lithium-ion battery chemistries, such as layered oxides cathodes NMC and LCO. Due to its low cost, low toxicity, stability, and reliable performance, LMFP-based batteries are increasingly applied in vehicle and backup power applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service