Key Documents
BR707525
BRAND® titrating tube with screw cap for Titrette® 10 mL
with integrated discharge/recircirculation valve
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, Dispensette®
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
BRAND 707525
General description
The BRAND® titrating tube with screw cap is specifically designed for use with the Titrette® 10 mL titrators. This tube provides a secure and reliable solution for conducting titrations, ensuring accurate measurements and ease of use. The screw cap ensures a tight seal, preventing leaks and contamination during the titration process.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)