GF91051501
Bismuth
lump, 100 mm max. lump size, weight 1126 g, purity 99.999%
Synonym(s):
BI006115
About This Item
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
99.999%
form
lumps
manufacturer/tradename
Goodfellow 910-515-01
resistivity
129 μΩ-cm, 20°C
weight
1126 g
max. lump size
100 mm
bp
1560 °C (lit.)
mp
271 °C (lit.)
density
9.8 g/mL at 25 °C (lit.)
SMILES string
[Bi]
InChI
1S/Bi
InChI key
JCXGWMGPZLAOME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service