Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10728

Sigma-Aldrich

m-dPEG®36-OH

>90% (HPLC)

Synonym(s):

m-PEG36-OH, mPEG-alcohol, mPEG1500, mPEG36-OH, mPEG36-alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H148O37
Molecular Weight:
1617.93
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

>90% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI key

BYLIGSNYHOODII-UHFFFAOYSA-N

Features and Benefits

m-dPEG36-OH is a chemical modification reagent. It consists of a methyl-terminated, water-soluble, non-immunogenic, single molecular weight, discrete polyethylene glycol (dPEG) chain. The dPEG chain is 109 atoms (130.1 Å) long and terminates with a reactive hydroxy group. The hydroxy group can be functionalized with different groups that enable various types of reactions. Uses for this monodisperse equivalent of an m-PEG1500-OH include surface passivation and biomolecule modification.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service