Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860567P

Avanti

18:1-6:0 Biotin PI(4,5)P2

Avanti Research - A Croda Brand 860567P, powder

Synonym(s):

1-oleoyl-2-[6-biotinyl(aminohexanoyl)]-sn-glycero-3-phosphoinositol-4,5-bisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H87N6O21P3S
CAS Number:
Molecular Weight:
1149.16
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (860567P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (860567P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 860567P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](OP(O)([O-])=O)[C@@H](O)[C@H]1O)([O-])=O)(OC(CCCCCNC(CCCC[C@]2([H])SC[C@@]3([H])[C@]2([H])NC(N3)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

InChI key

FAWBNQDQNLXDOW-LRZFZKCMSA-N

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860567P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (860567P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service