Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

880237P

Avanti

DMG-PEG(2000)-Azide

new

Avanti Research - A Croda Brand

Synonym(s):

1,2-dimyristoyl-rac-glycero-3-(azidopolyethylene glycol-2000)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C121H239N3O49
CAS Number:
Molecular Weight:
2520.18
UNSPSC Code:
12352200

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (880237P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(=O)OCC(COCCOCCN=[N+]=[N-])OC(=O)CCCCCCCCCCCCC

InChI

1S/C35H67N3O6/c1-3-5-7-9-11-13-15-17-19-21-23-25-34(39)43-32-33(31-42-30-29-41-28-27-37-38-36)44-35(40)26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3

InChI key

FBHUXNGXIPXVAG-UHFFFAOYSA-N

General description

A challenge associated with LNP′s is effectively delivering to the target cells. However, specific functional groups can be used to target these desirable cells. Click chemistry presents a promising solution for the straightforward attachment of functional groups to other molecules, enabling the synthesis of larger and more complex molecular structures. Clickable PEG lipids allow for the easy linkage of specific cell-targeting functional groups to the PEG lipid, enhancing delivery to the intended cells. Additionally, by adjusting the PEG length, one can modify micelle size, immunological safety, and drug release rates.

Application

Targeted drug delivery

Packaging

5 mL Amber Glass Screw Cap Vial (880242P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service