Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-0179-00

SMC® Plate-Based Assay Development Kit (5 Plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

technique(s)

single molecule counting: suitable

General description

Take advantage of Single Molecule Counting (SMC) technology while developing your own application-specific immunoassays in a plate-based format. This all-in-one kit provides everything you need to build your own high performing SMC immunoassay.

Application

Develop assays for either the TM=["SMCxPRO"] or Erenna

Components

Detection Label Buffer 1 Buffer 2 Buffer 3 (10X) Filter Tube Ultra 4 Assay Buffer Blocking Buffer Plate Standard Diluent Elution Buffer B Buffer C 10X PBS 10X Wash Buffer 10X Systems Buffer w/ 0.5% ProClin Optimization Standard Diluent A Optimization Standard Diluent B Optimization Standard Diluent C

Legal Information

ProClin is a trademark of LANXESS Corp.
SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service