Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04-1658

Sigma-Aldrich

Rabbit Anti-Human Ig light chain Antibody, clone RM129, biotin conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

RM129
monoclonal

purified by

using Protein A

species reactivity

rabbit, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable

isotype

IgG

General description

Human IgG

Specificity

This antibody reacts to both kappa and lambda light chains of human immunoglobulins. No cross reactivity with mouse, rat, or goat immunoglobulin light chain.

Quality

ELISA Analysis: The specificity and reactivity of a representative lot was confirmed by ELISA analyses.

Physical form

Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service