Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-972

Sigma-Aldrich

Anti-phospho-HP1 γ (Ser83) Antibody

Upstate®, from rabbit

Synonym(s):

Chromobox protein homolog 3, Heterochromatin protein 1 homolog gamma, Modifier 2 protein, HECH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer83)

Gene Information

human ... CBX3(11335)

Specificity

HP1γ phosphorylated at Ser83.

Immunogen

Phosphopeptide corresponding to amino acid position 76-88 (DGTKRKpSLSDSE-88) of human HP1 gamma.

Application

Anti-phospho-HP1 γ (Ser83) Antibody is a Rabbit Polyclonal Antibody for detection of phospho-HP1 γ (Ser83) also known as Heterochromatin protein 1 homolog gamma, HECH & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Routinely evaluated by immunoblot.

Target description

20 kDa

Physical form

Format: Purified
ImmunoAffinity Purified
Immunoaffinity purified rabbit IgG in PBS containing 30% glycerol.

Storage and Stability

2 years at -20°C from date of shipment.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service