Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-450-D

Sigma-Aldrich

CDK1/CyclinB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.27

Assay

≥75%

Quality Level

specific activity

90 pmol/min-μg

concentration

0.1 mg/mL

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... CDC2(983)

General description

FUNCTION: SwissProt: P06493 # Plays a key role in the control of the eukaryotic cell cycle. It is required in higher cells for entry into S-phase and mitosis. p34 is a component of the kinase complex that phosphorylates the repetitive C-terminus of RNA polymerase II.
SIZE: 297 amino acids; 34095 Da
SUBUNIT: Forms a stable but non-covalent complex with a regulatory subunit and with a cyclin. Interacts with DLG7. Isoform 2 is unable to complex with cyclin B1 and also fails to bind to the CDK inhibitor p21. Interacts with catalytically active CCNB1 and RALBP1 during mitosis to form an endocytotic complex during interphase.
SUBCELLULAR LOCATION: Nucleus (By similarity).
SIMILARITY: SwissProt: P06493 ## Belongs to the protein kinase superfamily. CMGC Ser/Thr protein kinase family. CDC2/CDKX subfamily. & Contains 1 protein kinase domain.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-450M-D when ordering the 250 µg size.

Quality

Evaluated by Kinase assay: 10 µl of CDK1/CyclinB1 was mixed with 5 µl Histone H1 substrate (1 mg/ml) and 5 µl water. Reaction was initiated by adding 5 µl of [33P]-ATP cocktail and incubated at 30°C for 15 min. Reaction was read by adding 20 µl to P81 paper and read in the presence of scintillation fluid in a scintillation counter.

Physical properties

CDK1: 58 kDa, CyclinB1: 75 kDa

Physical form

50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol

Storage and Stability

Stable for > 6 month at –80°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service