Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-055

Sigma-Aldrich

Protease Substrate Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

technique(s)

activity assay: suitable

Quality Level

General description

Millipore′s ready-to-screen Upstate Protease Substrate Set is comprised of a total of 360 peptides derived from cleavage sites described in the scientific literature. The Protease Substrate Set comes in a 384-well microtiter plate (75 pmol per well). In addition, several empty wells are available for positive and negative controls (wells P1-P24). Subsequent to incubation with the target protease, evolving fluorescence can be detected using standard microtiter plate readers. The sensitivity of the assay is sufficient to detect low nanogram levels of protease. The peptide derivatives are purified by HPLC (>95% at 220 nm) and freeze-dried into the wells from a DMSO stock solution. There are no additional buffer salts inside the wells. The peptide derivatives are not immobilized onto the walls of the wells resulting in a homogeneous assay after dissolving of the peptides in assay buffer.
The data CD-ROM provided with the set contains all information needed for the detailed analysis of your data including peptide sequences and Swiss-Prot accession numbers for the proteins containing these cleavage sites. If the individual human peptide sequence could be found in other organisms, too, all appropriate Swiss-Prot accession numbers are given.

Storage and Stability

Protease Substrate Set microtiter plates should be stored at -20°C. All other components may be stored at room temperature.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service