Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

419820

Sigma-Aldrich

BMS-509744

≥97% (HPLC), powder, ITK inhibitor, Calbiochem®

Synonym(s):

ITK Inhibitor, BMS-509744

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H41N5O4S2
CAS Number:
Molecular Weight:
623.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

ITK Inhibitor, BMS-509744,

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[s]1c(ncc1Sc3c(cc(c(c3)C(=O)N4CCN(CC4)C(=O)C)OC)C)NC(=O)c2ccc(cc2)CNC(C(C)(C)C)C

InChI

1S/C32H41N5O4S2/c1-20-16-26(41-7)25(30(40)37-14-12-36(13-15-37)22(3)38)17-27(20)42-28-19-34-31(43-28)35-29(39)24-10-8-23(9-11-24)18-33-21(2)32(4,5)6/h8-11,16-17,19,21,33H,12-15,18H2,1-7H3,(H,34,35,39)

InChI key

ZHXNIYGJAOPMSO-UHFFFAOYSA-N

General description

A cell-permeable aminothioaryl-thiazolo compound that potently inhibits ITK kinase activity (IC50 = 19 nM; [ATP] = 1 M) in an ATP-competitive manner by stabilizing ITK activation loop in a substrate-blocking, inactive conformation, inhibiting Fyn, IR, Lck, Btk only at much higher concentrations (IC50 = 1.1, 1.1, 2.4, and 4.1 M, respectively) and exhibiting little or no activity against 14 other kinases (IC50 ≥11 M). Inhibits ITK-dependent, but not ITK-independent, PLCγ1 tyrosine phosphorylation (IC50<300 nM in αCD3-stimulated Jurkat T-cells) as well as downstream signaling events, including Ca2+ mobilization (IC50 = 52 nM in αCD3-stimulated Jurkat T-cells), IL-2 secretion (IC50 = 72, 250, 380, and 390 nM, respectively, in αCD3-stimulated EL4, Jurkat, murine splenocytes, and human PBMCs), and proliferation (IC50 = 430 nM against APC-induced primary human T-cell expansion). BMS-509744 is also efficacious in alleviating OVA (Cat. No. 32467) challenge-induced airway leukocytes infiltration (50% and 75% reduction, respectively, of total and eosinophil cell count; 25 mg/kg s.c.) in OVA- sensitized mice in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ITK
Target IC50: 19 nM for ITK

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kutach, A.K., et al. 2010. Chem. Biol. Drug Des.76, 154.
Das, J., et al. 2006. Bioorg. Med. Chem. Lett.16, 3706.
Lin, T.A., et al. 2004. Biochemistry43, 11056.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service